Anti-HBsAg Mouse monoclonal antibody

Supplier: Biorbyt
BIRBORB24070-1EA 0 EUR
BIRBORB24070-1
Anti-HBsAg Mouse monoclonal antibody
Antibodies
Type: Primary
Antigen: HBsAg
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR