Anti-Borrelia burgdorferi sensu stric Mouse monoclonal antibody

Supplier: Biorbyt
BIRBORB24111-1EA 0 EUR
BIRBORB24111-1
Anti-Borrelia burgdorferi sensu stric Mouse monoclonal antibody
Antibodies
Type: Primary
Antigen: Borrelia burgdorferi sensu stric
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR