Anti-HIV-1 p17 Mouse monoclonal antibody [clone: 4A1]

Supplier: Biorbyt
BIRBORB24123-100EA 0 EUR
BIRBORB24123-100
Anti-HIV-1 p17 Mouse monoclonal antibody [clone: 4A1]
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: HIV-1 p17
Clonality: monoclonal
Clone: 4A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR