Anti-HSV-2 gB Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24137-100EA 0 EUR
BIRBORB24137-100
Anti-HSV-2 gB Mouse Monoclonal Antibody
Antibodies
ELISA: 1:10000; Western Blot: 1:1000; Immunofluorescence: 1:1000

Type: Primary
Antigen: HSV-2 gB
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR