Anti-RLN2 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24317-10EA 0 EUR
BIRBORB24317-10
Anti-RLN2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: RLN2 (relaxin 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR