Imprimer

Anti-TSH intact Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB24375-1EA 0 EUR
BIRBORB24375-1
Anti-TSH intact Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TSH intact
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR