Anti-Triiodothyronine Mouse Monoclonal Antibody [clone: 3A6]

Supplier: Biorbyt
BIRBORB24392-1EA 0 EUR
BIRBORB24392-1
Anti-Triiodothyronine Mouse Monoclonal Antibody [clone: 3A6]
Antibodies
Type: Primary
Antigen: Triiodothyronine
Clonality: monoclonal
Clone: 3A6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR