Anti-GHRL Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24424-1EA 0 EUR
BIRBORB24424-1
Anti-GHRL Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: GHRL (ghrelin/obestatin prepropeptide)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR