Anti-E. coli O157:H7 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB24609-1EA 0 EUR
BIRBORB24609-1
Anti-E. coli O157:H7 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: E. coli O157:H7
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR