Anti-MMP3 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24689-100EA 0 EUR
BIRBORB24689-100
Anti-MMP3 Mouse Monoclonal Antibody
Antibodies
Immunochemistry: 2-5 ?g/ml; ELISA: 1:3000-1:8000

Type: Primary
Antigen: MMP3 (matrix metallopeptidase 3 (stromelysin 1, progelatinase))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR