Anti-proANP Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24704-10EA 0 EUR
BIRBORB24704-10
Anti-proANP Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: proANP
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR