Anti-LDL Receptor Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24721-50EA 0 EUR
BIRBORB24721-50
Anti-LDL Receptor Mouse Monoclonal Antibody
Antibodies
  • Immunofluorescence: 1:10-1:50

Type: Primary
Antigen: LDL Receptor
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR