Anti-Alpha Fetoprotein Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24736-1EA 0 EUR
BIRBORB24736-1
Anti-Alpha Fetoprotein Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Alpha Fetoprotein
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR