Anti-LEP Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24760-100EA 0 EUR
BIRBORB24760-100
Anti-LEP Mouse Monoclonal Antibody
Antibodies
  • Western Blot: 1:4000

Type: Primary
Antigen: LEP (leptin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR