Anti-Low Density LRP (85kDa) Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24807-100EA 0 EUR
BIRBORB24807-100
Anti-Low Density LRP (85kDa) Mouse Monoclonal Antibody
Antibodies
ELISA: 1:10000; Western Blot: 1-5 ?g/ml; Immunofluorescence: 1-10 ?g/ml

Type: Primary
Antigen: Low Density LRP (85kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR