Anti-GAPDH Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Biorbyt
BIRBORB24833-1EA 0 EUR
BIRBORB24833-1
Anti-GAPDH Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR