Anti-HSP-70, M. tuberculosis Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24921-1EA 0 EUR
BIRBORB24921-1
Anti-HSP-70, M. tuberculosis Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: HSP-70, M. tuberculosis
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR