Anti-ACSM3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25094-1EA 0 EUR
BIRBORB25094-1
Anti-ACSM3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ACSM3 (acyl-CoA synthetase medium-chain family member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR