Anti-V5 Tag Chicken Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25120-100EA 0 EUR
BIRBORB25120-100
Anti-V5 Tag Chicken Polyclonal Antibody
Antibodies
ELISA: 1:100000; Western Blot: 1:5000

Type: Primary
Antigen: V5 Tag (V5 tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR