Anti-APOA2 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25143-1EA 0 EUR
BIRBORB25143-1
Anti-APOA2 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: APOA2 (apolipoprotein A-II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR