Imprimer

Anti-Scl-70 Ag Human Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25154-2EA 0 EUR
BIRBORB25154-2
Anti-Scl-70 Ag Human Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Scl-70 Ag
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Human
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR