Anti-Positive Control Set For Autoimmune Human Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25157-1EA 0 EUR
BIRBORB25157-1
Anti-Positive Control Set For Autoimmune Human Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Positive Control Set For Autoimmune
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Human
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR