Anti-NPR-Bi Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25188-20EA 0 EUR
BIRBORB25188-20
Anti-NPR-Bi Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100; Immunochemistry: 1:100

Type: Primary
Antigen: NPR-Bi
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR