Anti-PSAT1 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25247-1EA 0 EUR
BIRBORB25247-1
Anti-PSAT1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PSAT1 (phosphoserine aminotransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR