Anti-APOB Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25283-1EA 0 EUR
BIRBORB25283-1
Anti-APOB Goat Polyclonal Antibody
Antibodies
DB: 1:200-1:10000

Type: Primary
Antigen: APOB (apolipoprotein B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR