Imprimer

Anti-MPO Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25303-1EA 0 EUR
BIRBORB25303-1
Anti-MPO Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:4000

Type: Primary
Antigen: MPO (myeloperoxidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR