Anti-NF-Y Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25347-100EA 0 EUR
BIRBORB25347-100
Anti-NF-Y Rabbit Polyclonal Antibody
Antibodies
DB: 1:1000

Type: Primary
Antigen: NF-Y
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR