Anti-Luciferase Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Biorbyt
BIRBORB25418-20EA 0 EUR
BIRBORB25418-20
Anti-Luciferase Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
ELISA: 1:2000-1:12000

Type: Primary
Antigen: Luciferase
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR