Anti-PARC Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25442-100EA 0 EUR
BIRBORB25442-100
Anti-PARC Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:2000-1:10000; Western Blot: 1:500-1:1000

Type: Primary
Antigen: PARC
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR