Anti-APOE Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25475-10EA 0 EUR
BIRBORB25475-10
Anti-APOE Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR