Anti-Rat Renin Sheep Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25481-1EA 0 EUR
BIRBORB25481-1
Anti-Rat Renin Sheep Polyclonal Antibody
Antibodies
ELISA: 1:50000; Western Blot: 1:5000; Immunofluorescence: 1:1000

Type: Primary
Antigen: Rat Renin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR