Anti-Phosphoserine Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB25508-100EA 0 EUR
BIRBORB25508-100
Anti-Phosphoserine Mouse Monoclonal Antibody
Antibodies
Western Blot: 1 ?g/ml; ELISA: 0.05 ?g/ml

Type: Primary
Antigen: Phosphoserine
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR