Anti-GAPDH Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25688-100EA 0 EUR
BIRBORB25688-100
Anti-GAPDH Rabbit Polyclonal Antibody
Antibodies
Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR