Anti-Glyco.Synthase Kin.3b Mouse monoclonal antibody

Supplier: Biorbyt
BIRBORB25698-100EA 0 EUR
BIRBORB25698-100
Anti-Glyco.Synthase Kin.3b Mouse monoclonal antibody
Antibodies
  • Western Blot: 1:100; ELISA: 1:100

Type: Primary
Antigen: Glyco.Synthase Kin.3b
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR