Anti-C1QBP Sheep Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25702-1EA 0 EUR
BIRBORB25702-1
Anti-C1QBP Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: C1QBP (complement component 1, q subcomponent binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR