Anti-ALB Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Biorbyt
BIRBORB25713-1EA 0 EUR
BIRBORB25713-1
Anti-ALB Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
EIA: 1:12800-1:25600

Type: Primary
Antigen: ALB (albumin)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR