Anti-VWF Sheep Polyclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
BIRBORB25724-1EA 0 EUR
BIRBORB25724-1
Anti-VWF Sheep Polyclonal Antibody (FITC (Fluorescein))
Antibodies
Immunofluorescence: 1:50-1:100

Type: Primary
Antigen: VWF (von Willebrand factor)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR