Anti-ERBB2 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB25904-1EA 0 EUR
BIRBORB25904-1
Anti-ERBB2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ERBB2 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR