Anti-CKM Mouse monoclonal antibody

Supplier: Biorbyt
BIRBORB26047-1EA 0 EUR
BIRBORB26047-1
Anti-CKM Mouse monoclonal antibody
Antibodies
Type: Primary
Antigen: CKM (creatine kinase, muscle)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR