Anti-MME Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB26101-100EA 0 EUR
BIRBORB26101-100
Anti-MME Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: MME (membrane metallo-endopeptidase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR