Anti-FCGR3A Mouse Monoclonal Antibody (Biotin)

Supplier: Biorbyt
BIRBORB26106-100EA 0 EUR
BIRBORB26106-100
Anti-FCGR3A Mouse Monoclonal Antibody (Biotin)
Antibodies
Flow Cytometry: 1:500

Type: Primary
Antigen: FCGR3A (Fc fragment of IgG, low affinity IIIa, receptor (CD16a))
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR