Anti-NCAM1 Mouse Monoclonal Antibody (Biotin)

Supplier: Biorbyt
BIRBORB26122-100EA 0 EUR
BIRBORB26122-100
Anti-NCAM1 Mouse Monoclonal Antibody (Biotin)
Antibodies
Flow Cytometry: 1:200

Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR