Anti-CD44 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB26130-0.2EA 0 EUR
BIRBORB26130-0.2
Anti-CD44 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR