Anti-CEACAM5 Mouse monoclonal antibody

Supplier: Biorbyt
BIRBORB26143-1EA 0 EUR
BIRBORB26143-1
Anti-CEACAM5 Mouse monoclonal antibody
Antibodies
Type: Primary
Antigen: CEACAM5 (carcinoembryonic antigen-related cell adhesion molecule 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR