Anti-E. coli SLT-1 (STX-1) Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB26201-1EA 0 EUR
BIRBORB26201-1
Anti-E. coli SLT-1 (STX-1) Mouse Monoclonal Antibody
Antibodies
  • DB: 1 μg/ml; ELISA: 100 μl

Type: Primary
Antigen: E. coli SLT-1 (STX-1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR