Imprimer

Anti-S100 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB26221-100EA 0 EUR
BIRBORB26221-100
Anti-S100 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: S100
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR