Anti-Mouse Plasminogen Rat Monoclonal Antibody

Supplier: Biorbyt
BIRBORB26252-100EA 0 EUR
BIRBORB26252-100
Anti-Mouse Plasminogen Rat Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Mouse Plasminogen
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR