Anti-CD13 Myeloid Cells Mouse Monoclonal Antibody (Biotin)

Supplier: Biorbyt
BIRBORB26355-0.2EA 0 EUR
BIRBORB26355-0.2
Anti-CD13 Myeloid Cells Mouse Monoclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CD13 Myeloid Cells
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR