Imprimer

Anti-CR1 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB26502-0.2EA 0 EUR
BIRBORB26502-0.2
Anti-CR1 Mouse Monoclonal Antibody
Anticorps
  • Flow Cytometry: 2 μg; Western Blot: 1:25-1:50

Type: Primary
Antigen: CR1 (complement component (3b/4b) receptor 1 (Knops blood group))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR