Imprimer

Anti-CD86 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB26559-0.2EA 0 EUR
BIRBORB26559-0.2
Anti-CD86 Mouse Monoclonal Antibody
Anticorps
Flow Cytometry: 1:20-1:100; Immunochemistry: 1:20-1:100

Type: Primary
Antigen: CD86 (CD86 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR